accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 39786-35-1

IUPAC Name: ethyl 3-amino-1-benzofuran-2-carboxylate
Synonyms: ethyl 3-amino-1-benzofuran-2-carboxylate
Molecular Weight (g/mol): 205.21
Molecular Formula: C11H11NO3
InChi Key: PWOARNMOPCOJEV-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(N)C2=CC=CC=C2O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results