accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 398-62-9

H3COFOCH3

CAS RN 398-62-9

IUPAC Name: 4-fluoro-1,2-dimethoxybenzene
Synonyms: 4-fluoro-1,2-dimethoxybenzene
Molecular Weight (g/mol): 156.16
Molecular Formula: C8H9FO2
InChi Key: DAGKHJDZYJFWSO-UHFFFAOYSA-N
SMILES: COC1=CC=C(F)C=C1OC
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)