accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNHOCH3

CAS RN 39887-40-6

IUPAC Name: 4-methyl-N-propylbenzamide
Synonyms: 4-methyl-N-propylbenzamide
Molecular Weight (g/mol): 177.25
Molecular Formula: C11H15NO
InChi Key: MEKGGTFUDJLKHR-UHFFFAOYSA-N
SMILES: CCCNC(=O)C1=CC=C(C)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results