accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOSNN

CAS RN 3989-36-4

IUPAC Name: ethyl 1,2,3-thiadiazole-4-carboxylate
Synonyms: ethyl 1,2,3-thiadiazole-4-carboxylate
Molecular Weight (g/mol): 158.18
Molecular Formula: C5H6N2O2S
InChi Key: FBUHTNOJXVICFM-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CSN=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results