accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 399-30-4

CH3H2NF

CAS RN 399-30-4

IUPAC Name: [(2-fluorophenyl)methyl](methyl)azanium
Synonyms: [(2-fluorophenyl)methyl](methyl)azanium
Molecular Weight (g/mol): 140.18
Molecular Formula: C8H11FN
InChi Key: AHIHZCXUWGORQO-UHFFFAOYSA-O
SMILES: C[NH2+]CC1=CC=CC=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results