accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 399-51-9

IUPAC Name: 6-fluoro-1H-indole
Synonyms: 6-fluoro-1H-indole
Molecular Weight (g/mol): 135.14
Molecular Formula: C8H6FN
InChi Key: YYFFEPUCAKVRJX-UHFFFAOYSA-N
SMILES: FC1=CC=C2C=CNC2=C1
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results