accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2FHO

CAS RN 399-97-3

IUPAC Name: 2-amino-4-fluorophenol
Synonyms: 2-amino-4-fluorophenol
Molecular Weight (g/mol): 127.12
Molecular Formula: C6H6FNO
InChi Key: ULDFRPKVIZMKJG-UHFFFAOYSA-N
SMILES: NC1=CC(F)=CC=C1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results