accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 400-76-0

IUPAC Name: 4-amino-3-(trifluoromethyl)benzoic acid
Synonyms: 4-amino-3-(trifluoromethyl)benzoic acid
Molecular Weight (g/mol): 205.14
Molecular Formula: C8H6F3NO2
InChi Key: NPPPORJZPNJXNQ-UHFFFAOYSA-N
SMILES: NC1=C(C=C(C=C1)C(O)=O)C(F)(F)F
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results