accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 4002-83-9

IUPAC Name: 1-methyl-1H-indazole-3-carbaldehyde
Synonyms: 1-methylindazole-3-carbaldehyde
Molecular Weight (g/mol): 160.18
Molecular Formula: C9H8N2O
InChi Key: KYDLGYCLCFKUHY-UHFFFAOYSA-N
SMILES: CN1N=C(C=O)C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results