accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BOHHO(?)

CAS RN 400607-46-7

IUPAC Name: [10-(naphthalen-1-yl)anthracen-9-yl]boronic acid
Synonyms: 10-(naphthalen-1-yl)anthracen-9-ylboronic acid
Molecular Weight (g/mol): 348.21
Molecular Formula: C24H17BO2
InChi Key: ASQXKNXJNDLXQV-UHFFFAOYSA-N
SMILES: OB(O)C1=C2C=CC=CC2=C(C2=CC=CC=C12)C1=C2C=CC=CC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results