accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NSSOHO

CAS RN 400715-45-9

IUPAC Name: 5-(2-methyl-1,3-thiazol-4-yl)thiophene-2-carboxylic acid
Synonyms: 5-(2-methyl-1,3-thiazol-4-yl)thiophene-2-carboxylic acid
Molecular Weight (g/mol): 225.28
Molecular Formula: C9H7NO2S2
InChi Key: AEDWMWQOFJYAGN-UHFFFAOYSA-N
SMILES: CC1=NC(=CS1)C1=CC=C(S1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results