accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BrBrFFF

CAS RN 401-84-3

IUPAC Name: 1,3-dibromo-5-(trifluoromethyl)benzene
Synonyms: 1,3-dibromo-5-(trifluoromethyl)benzene
Molecular Weight (g/mol): 303.9
Molecular Formula: C7H3Br2F3
InChi Key: OFNXSUANJLHGQN-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(Br)=CC(Br)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results