accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 40106-13-6

H3COOH2NS

CAS RN 40106-13-6

IUPAC Name: ethyl 2-amino-4H,5H,6H,7H,8H-cyclohepta[b]thiophene-3-carboxylate
Synonyms: ethyl 2-amino-4H,5H,6H,7H,8H-cyclohepta[b]thiophene-3-carboxylate
Molecular Weight (g/mol): 239.33
Molecular Formula: C12H17NO2S
InChi Key: XTUHIGALMIGZST-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(N)SC2=C1CCCCC2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results