accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOHNOHHCl

CAS RN 40126-30-5

IUPAC Name: methyl 4-hydroxypyrrolidine-2-carboxylate hydrochloride
Synonyms: methyl 4-hydroxypyrrolidine-2-carboxylate hydrochloride
Molecular Weight (g/mol): 181.62
Molecular Formula: C6H12ClNO3
InChi Key: KLGSHNXEUZOKHH-UHFFFAOYNA-N
SMILES: Cl.COC(=O)C1CC(O)CN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results