accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 4018-82-0

H3COHNOCl

CAS RN 4018-82-0

IUPAC Name: 4-chloro-N-(4-methoxyphenyl)benzamide
Synonyms: 4-chloro-N-(4-methoxyphenyl)benzamide
Molecular Weight (g/mol): 261.71
Molecular Formula: C14H12ClNO2
InChi Key: UVGPQLMHIRNFEE-UHFFFAOYSA-N
SMILES: COC1=CC=C(NC(=O)C2=CC=C(Cl)C=C2)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results