accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FFFBr

CAS RN 402-23-3

IUPAC Name: 1-(bromomethyl)-3-(trifluoromethyl)benzene
Synonyms: 1-(bromomethyl)-3-(trifluoromethyl)benzene
Molecular Weight (g/mol): 239.04
Molecular Formula: C8H6BrF3
InChi Key: MYYYZNVAUZVXBO-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=CC(CBr)=C1
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results