accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 402-43-7

IUPAC Name: 1-bromo-4-(trifluoromethyl)benzene
Synonyms: 1-bromo-4-(trifluoromethyl)benzene
Molecular Weight (g/mol): 225.01
Molecular Formula: C7H4BrF3
InChi Key: XLQSXGGDTHANLN-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=C(Br)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)