accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 402-45-9

OHFFF

CAS RN 402-45-9

IUPAC Name: 4-(trifluoromethyl)phenol
Synonyms: α,α,α-trifluoro-P-cresol P-trifluoromethylphenol α,α,α-trifluoro-p-cresol
Molecular Weight (g/mol): 162.11
Molecular Formula: C7H5F3O
InChi Key: BAYGVMXZJBFEMB-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)C(F)(F)F
Certifications/Compliance 
  • (3)
  • (1)
Boiling Point 
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results