accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 402-61-9

CH3NHNOHO

CAS RN 402-61-9

IUPAC Name: 5-methyl-1H-pyrazole-3-carboxylic acid
Synonyms: 5-methyl-1H-pyrazole-3-carboxylic acid
Molecular Weight (g/mol): 126.12
Molecular Formula: C5H6N2O2
InChi Key: WSMQKESQZFQMFW-UHFFFAOYSA-N
SMILES: CC1=CC(=NN1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results