CAS RN 402-67-5
IUPAC Name:
1-fluoro-3-nitrobenzene
Synonyms:
3-fluoronitrobenzene
Molecular Weight (g/mol):
141.1
Molecular Formula:
C6H4FNO2
InChi Key:
WMASLRCNNKMRFP-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=CC=CC(F)=C1
1
–
1
of
1
results
1
1-Fluoro-3-nitrobenzene, 98+%
CAS: 402-67-5 Molecular Formula: C6H4FNO2 Molecular Weight (g/mol): 141.10 MDL Number: MFCD00007196 InChI Key: WMASLRCNNKMRFP-UHFFFAOYSA-N Synonym: 3-fluoronitrobenzene,m-fluoronitrobenzene,m-nitrofluorobenzene,benzene, 1-fluoro-3-nitro,3-fluoro-1-nitrobenzene,1-fluoro-3-nitro-benzene,3-fluoro-nitrobenzene,3-nitro-fluorobenzene,pubchem2286,3-fluoro nitro benzene PubChem CID: 9823 IUPAC Name: 1-fluoro-3-nitrobenzene SMILES: [O-][N+](=O)C1=CC=CC(F)=C1