CAS RN 40210-84-2
IUPAC Name:
2-(2H-1,3-dithiol-2-ylidene)-2H-1,3-dithiole; 2-[4-(dicyanomethylidene)cyclohexa-2,5-dien-1-ylidene]propanedinitrile
Synonyms:
tetracyanoquinodimethane; tetrathiafulvalene
Molecular Weight (g/mol):
408.53
Molecular Formula:
C18H8N4S4
InChi Key:
OIXMVDHMELKBDX-UHFFFAOYSA-N
SMILES:
S1C=CSC1=C1SC=CS1.N#CC(C#N)=C1C=CC(C=C1)=C(C#N)C#N
1
–
1
of
1
results
1
Tetrathiafulvalene - 7,7,8,8-Tetracyanoquinodimethane Complex, TRC
CAS: 40210-84-2 Molecular Formula: C6H4S4 . C12H4N4 Molecular Weight (g/mol): 408.54 IUPAC Name: 2-[4-(dicyanomethylidene)cyclohexa-2,5-dien-1-ylidene]propanedinitrile;2-(1,3-dithiol-2-ylidene)-1,3-dithiole SMILES: N#CC(=C1C=CC(=C(C#N)C#N)C=C1)C#N.S2C=CSC2=C3SC=CS3