accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NPNN

CAS RN 4023-53-4

IUPAC Name: 3-[bis(2-cyanoethyl)phosphanyl]propanenitrile
Synonyms: 3-[bis(2-cyanoethyl)phosphanyl]propanenitrile
Molecular Weight (g/mol): 193.19
Molecular Formula: C9H12N3P
InChi Key: CHZAMJVESILJGH-UHFFFAOYSA-N
SMILES: N#CCCP(CCC#N)CCC#N
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)