accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 40244-90-4

IUPAC Name: chlorobis(propan-2-yl)phosphane
Synonyms: chlorodiisopropylphosphine
Molecular Weight (g/mol): 152.6
Molecular Formula: C6H14ClP
InChi Key: JZPDBTOWHLZQFC-UHFFFAOYSA-N
SMILES: CC(C)P(Cl)C(C)C
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results