accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONOSClOO

CAS RN 4025-75-6

IUPAC Name: (4-nitrophenyl)methanesulfonyl chloride
Synonyms: (4-nitrophenyl)methanesulfonyl chloride
Molecular Weight (g/mol): 235.64
Molecular Formula: C7H6ClNO4S
InChi Key: CAXRPEAFHWDFTD-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(CS(Cl)(=O)=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results