CAS RN 402615-91-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
rac FTY720 Phosphate, TRC
CAS: 402615-91-2 Molecular Formula: C19 H34 N O5 P Molecular Weight (g/mol): 387.45 Synonym: 1,3-Propanediol, 2-amino-2-[2-(4-octylphenyl)ethyl]-, 1-(dihydrogen phosphate),1,3-Propanediol, 2-amino-2-[2-(4-octylphenyl)ethyl]-, mono(dihydrogen phosphate) (ester) (9CI),FTY 720 phosphate,FTY 720P,FTY-P,Fingolimod phosphate,Fingolimod-P,rac FTY720 Phosphate,Fingolimod O-phosphate IUPAC Name: [2-amino-2-(hydroxymethyl)-4-(4-octylphenyl)butyl] dihydrogen phosphate SMILES: CCCCCCCCc1ccc(CCC(N)(CO)COP(=O)(O)O)cc1