accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FN

CAS RN 403-54-3

IUPAC Name: 3-fluorobenzonitrile
Synonyms: benzonitrile, 3-fluoro-
Molecular Weight (g/mol): 121.11
Molecular Formula: C7H4FN
InChi Key: JZTPKAROPNTQQV-UHFFFAOYSA-N
SMILES: FC1=CC=CC(=C1)C#N
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results