CAS RN 403-90-7
IUPAC Name:
2-amino-3-(3-fluoro-4-hydroxyphenyl)propanoic acid
Synonyms:
pardinon fluorthyrin m-ftyr
Molecular Weight (g/mol):
199.18
Molecular Formula:
C9H10FNO3
InChi Key:
VIIAUOZUUGXERI-UHFFFAOYNA-N
SMILES:
NC(CC1=CC=C(O)C(F)=C1)C(O)=O
1
–
1
of
1
results
1
3-Fluoro-DL-tyrosine, 97%, Thermo Scientific Chemicals
CAS: 403-90-7 Molecular Formula: C9H10FNO3 Molecular Weight (g/mol): 199.18 MDL Number: MFCD00063075 InChI Key: VIIAUOZUUGXERI-UHFFFAOYNA-N Synonym: 3-fluoro-dl-tyrosine,2-amino-3-3-fluoro-4-hydroxyphenyl propanoic acid,pardinon,m-fluoro-dl-tyrosine,fluorthyrin,3-fluorotyrosin,3-fluorotyrosine,m-ftyr,dl-m-fluorotyrosine,dl-3-fluorotyrosine PubChem CID: 92100 ChEBI: CHEBI:32771 IUPAC Name: 2-amino-3-(3-fluoro-4-hydroxyphenyl)propanoic acid SMILES: NC(CC1=CC=C(O)C(F)=C1)C(O)=O