accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNNOO

CAS RN 40332-17-0

IUPAC Name: 2-(2-methyl-1H-1,3-benzodiazol-1-yl)acetate
Synonyms: 2-(2-methyl-1,3-benzodiazol-1-yl)acetate
Molecular Weight (g/mol): 189.2
Molecular Formula: C10H9N2O2
InChi Key: SMGBDFGKVGSCGB-UHFFFAOYSA-M
SMILES: CC1=NC2=CC=CC=C2N1CC([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results