accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OH(S)NHOOOO

CAS RN 40371-50-4

IUPAC Name: (2S)-4-{[(benzyloxy)carbonyl]amino}-2-hydroxybutanoate
Synonyms: (2S)-4-{[(benzyloxy)carbonyl]amino}-2-hydroxybutanoate
Molecular Weight (g/mol): 252.25
Molecular Formula: C12H14NO5
InChi Key: ULKOBRDRCYROKY-JTQLQIEISA-M
SMILES: O[C@@H](CCNC(=O)OCC1=CC=CC=C1)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results