accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COSiSSSSSiOCH3OCH3OCH3OCH3OCH3

CAS RN 40372-72-3

IUPAC Name: 4,4,15,15-tetraethoxy-3,16-dioxa-8,9,10,11-tetrathia-4,15-disilaoctadecane
Synonyms: bis(triethoxysilylpropyl)tetrasulfide
Molecular Weight (g/mol): 538.94
Molecular Formula: C18H42O6S4Si2
InChi Key: VTHOKNTVYKTUPI-UHFFFAOYSA-N
SMILES: CCO[Si](CCCSSSSCCC[Si](OCC)(OCC)OCC)(OCC)OCC
Boiling Point 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)