accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

LiH3CSiCH3H3CNSiCH3CH3H3C

CAS RN 4039-32-1

IUPAC Name: lithium(1+) bis(trimethylsilyl)azanide
Synonyms: lithium bis(trimethylsilyl)amide
Molecular Weight (g/mol): 167.33
Molecular Formula: C6H18LiNSi2
InChi Key: YNESATAKKCNGOF-UHFFFAOYSA-N
SMILES: [Li+].C[Si](C)(C)[N-][Si](C)(C)C
Molecular Weight (g/mol) 
  • (8)
  • (11)
Quantity 
  • (1)
  • (1)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
Physical Form 
  • (3)
  • (4)
  • (3)
  • (2)
  • (2)
Boiling Point 
  • (3)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (8)
  • (11)
  • (1)
  • (1)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
  • (3)
  • (4)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)