accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

SCNNCS

CAS RN 4044-65-9

IUPAC Name: 1,4-diisothiocyanatobenzene
Synonyms: bitoscanate
Molecular Weight (g/mol): 192.25
Molecular Formula: C8H4N2S2
InChi Key: OMWQUXGVXQELIX-UHFFFAOYSA-N
SMILES: S=C=NC1=CC=C(C=C1)N=C=S
Molecular Weight (g/mol) 
  • (3)
  • (2)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Certifications/Compliance 
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)