accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 4048-33-3

IUPAC Name: 6-aminohexan-1-ol
Synonyms: hexanol, 6-amino-
Molecular Weight (g/mol): 117.19
Molecular Formula: C6H15NO
InChi Key: SUTWPJHCRAITLU-UHFFFAOYSA-N
SMILES: NCCCCCCO
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (5)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)