CAS RN 406-58-6
IUPAC Name:
1,1,1,3,3-pentafluorobutane
Synonyms:
hfc-365mfc
Molecular Weight (g/mol):
148.08
Molecular Formula:
C4H5F5
InChi Key:
WZLFPVPRZGTCKP-UHFFFAOYSA-N
SMILES:
CC(F)(F)CC(F)(F)F
1
–
1
of
1
results
1
1,1,1,3,3-Pentafluorobutane, 99.5+%
CAS: 406-58-6 Molecular Formula: C4H5F5 Molecular Weight (g/mol): 148.076 MDL Number: MFCD03094115 InChI Key: WZLFPVPRZGTCKP-UHFFFAOYSA-N PubChem CID: 67884 IUPAC Name: 1,1,1,3,3-pentafluorobutane SMILES: CC(CC(F)(F)F)(F)F