accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOFFF

CAS RN 406-95-1

IUPAC Name: 2,2,2-trifluoroethyl acetate
Synonyms: trifluoroethyl acetate
Molecular Weight (g/mol): 142.08
Molecular Formula: C4H5F3O2
InChi Key: ZOWSJJBOQDKOHI-UHFFFAOYSA-N
SMILES: CC(=O)OCC(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results