accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3CH3H3CHNNHCH3CH3H3C

CAS RN 4062-60-6

IUPAC Name: tert-butyl[2-(tert-butylamino)ethyl]amine
Synonyms: tert-butyl[2-(tert-butylamino)ethyl]amine
Molecular Weight (g/mol): 172.32
Molecular Formula: C10H24N2
InChi Key: KGHYGBGIWLNFAV-UHFFFAOYSA-N
SMILES: CC(C)(C)NCCNC(C)(C)C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results