accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOH3CCH3OBr

CAS RN 40635-67-4

IUPAC Name: 1-bromo-2-methyl-1-oxopropan-2-yl acetate
Synonyms: 1-bromo-2-methyl-1-oxopropan-2-yl acetate
Molecular Weight (g/mol): 209.04
Molecular Formula: C6H9BrO3
InChi Key: OOKAXSHFTDPZHP-UHFFFAOYSA-N
SMILES: CC(=O)OC(C)(C)C(Br)=O
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)