accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 40649-36-3

CH3O

CAS RN 40649-36-3

IUPAC Name: 4-propylcyclohexan-1-one
Synonyms: 4-propylcyclohexan-1-one
Molecular Weight (g/mol): 140.23
Molecular Formula: C9H16O
InChi Key: NQEDLIZOPMNZMC-UHFFFAOYSA-N
SMILES: CCCC1CCC(=O)CC1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results