accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOHSOHOOO

CAS RN 4065-45-6

IUPAC Name: sodium 5-benzoyl-4-hydroxy-2-methoxybenzene-1-sulfonate
Synonyms: benzophenone-4
Molecular Weight (g/mol): 330.29
Molecular Formula: C14H11NaO6S
InChi Key: KJCLYACXIWMFCC-UHFFFAOYSA-M
SMILES: [Na+].COC1=CC(O)=C(C=C1S([O-])(=O)=O)C(=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results