accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 40675-81-8

IUPAC Name: 1-[(naphthalen-1-yl)methyl]piperazine-1,4-diium
Synonyms: 1-(naphthalen-1-ylmethyl)piperazine-1,4-diium
Molecular Weight (g/mol): 228.34
Molecular Formula: C15H20N2
InChi Key: HGYDREHWXXUUIS-UHFFFAOYSA-P
SMILES: C([NH+]1CC[NH2+]CC1)C1=C2C=CC=CC2=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results