accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2COOFFFFFFFFFFF

CAS RN 40677-94-9

IUPAC Name: (1,2,2,3,3,4,4,5,5,6,6-undecafluorocyclohexyl)methyl prop-2-enoate
Synonyms: (1,2,2,3,3,4,4,5,5,6,6-undecafluorocyclohexyl)methyl prop-2-enoate
Molecular Weight (g/mol): 366.13
Molecular Formula: C10H5F11O2
InChi Key: OZGWOALFBHODRB-UHFFFAOYSA-N
SMILES: FC1(F)C(F)(F)C(F)(F)C(F)(COC(=O)C=C)C(F)(F)C1(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results