accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

PPClClPdCH3CH3CH3H3CCH3H3C

CAS RN 40691-33-6

IUPAC Name: bis(tris(2-methylphenyl)phosphane) palladium dichloride
Synonyms: bis(tris(2-methylphenyl)phosphane) palladium dichloride
Molecular Weight (g/mol): 786.07
Molecular Formula: C42H42Cl2P2Pd
InChi Key: KFQYTZDYGOBBOE-UHFFFAOYSA-L
SMILES: [Cl-].[Cl-].[Pd].CC1=CC=CC=C1P(C1=CC=CC=C1C)C1=CC=CC=C1C.CC1=CC=CC=C1P(C1=CC=CC=C1C)C1=CC=CC=C1C
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results