accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FFFOOOFFF

CAS RN 407-25-0

IUPAC Name: 2,2,2-trifluoroacetyl 2,2,2-trifluoroacetate
Synonyms: trifluoroacetic anhydride
Molecular Weight (g/mol): 210.03
Molecular Formula: C4F6O3
InChi Key: QAEDZJGFFMLHHQ-UHFFFAOYSA-N
SMILES: FC(F)(F)C(=O)OC(=O)C(F)(F)F
Quantity 
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (4)
  • (6)
Percent Purity 
  • (6)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (4)
  • (4)
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
  • (6)
  • (6)
  • (3)
  • (6)