CAS RN 407-47-6
IUPAC Name:
2,2,2-trifluoroethyl prop-2-enoate
Synonyms:
2,2,2-trifluoroethyl prop-2-enoate
Molecular Weight (g/mol):
154.09
Molecular Formula:
C5H5F3O2
InChi Key:
VBHXIMACZBQHPX-UHFFFAOYSA-N
SMILES:
FC(F)(F)COC(=O)C=C
1
–
1
of
1
results
1
2,2,2-Trifluoroethyl acrylate, 98%, stab. with 200ppm 4-methoxyphenol
CAS: 407-47-6 Molecular Formula: C5H5F3O2 Molecular Weight (g/mol): 154.088 MDL Number: MFCD00000444 InChI Key: VBHXIMACZBQHPX-UHFFFAOYSA-N Synonym: 2,2,2-trifluoroethyl acrylate,2-propenoic acid, 2,2,2-trifluoroethyl ester,2-propenoic acid, trifluoroethyl ester,2,2,2-trifluoroethylacrylate,acrylic acid, 2,2,2-trifluoroethyl ester,acrylic acid 2,2,2-trifluoroethyl ester,tfol-a,acmc-20aokw,trifluoroethyl acrylate,pubchem12647 PubChem CID: 67889 IUPAC Name: 2,2,2-trifluoroethyl prop-2-enoate SMILES: C=CC(=O)OCC(F)(F)F