accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 407-97-6

IUPAC Name: 1-bromo-5-fluoropentane
Synonyms: 1-bromo-5-fluoropentane
Molecular Weight (g/mol): 169.04
Molecular Formula: C5H10BrF
InChi Key: GMYIZICPHREVDH-UHFFFAOYSA-N
SMILES: FCCCCCBr
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results