accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 4075-81-4

IUPAC Name: calcium dipropanoate
Synonyms: calcium propanoate
Molecular Weight (g/mol): 186.22
Molecular Formula: C6H10CaO4
InChi Key: BCZXFFBUYPCTSJ-UHFFFAOYSA-L
SMILES: [Ca++].CCC([O-])=O.CCC([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results