accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ICH3NCH3CH3CH3

CAS RN 40797-39-5

IUPAC Name: tetradodecylazanium iodide
Synonyms: tetradodecylazanium iodide
Molecular Weight (g/mol): 818.24
Molecular Formula: C48H100IN
InChi Key: OBMLOTDECOTAGU-UHFFFAOYSA-M
SMILES: [I-].CCCCCCCCCCCC[N+](CCCCCCCCCCCC)(CCCCCCCCCCCC)CCCCCCCCCCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results