accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 4084-27-9

HO(R)OH(S)OHOO

CAS RN 4084-27-9

IUPAC Name: (4S,5R)-4,5,6-trihydroxy-2-oxohexanal
Synonyms: 3-deoxyglucosone 3-deoxy-D-fructose 3-deoxy-erythro-hexos-2-ulose 3-deoxy-2-ketoglucose 3-deoxy-D-erythro-hexosulose 3-deoxy-D-glucosone 3-deoxy-D-erythro-hexulose
Molecular Weight (g/mol): 162.14
Molecular Formula: C6H10O5
InChi Key: ZGCHLOWZNKRZSN-NTSWFWBYSA-N
SMILES: OC[C@@H](O)[C@@H](O)CC(=O)C=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)