accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 4089-07-0

(S)ONH2OHOCH3HCl

CAS RN 4089-07-0

IUPAC Name: hydrogen ethyl 2-amino-3-(4-hydroxyphenyl)propanoate chloride
Synonyms: hydrogen L-tyrosine, ethyl ester chloride
Molecular Weight (g/mol): 245.7
Molecular Formula: C11H16ClNO3
InChi Key: BQULAXAVRFIAHN-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CCOC(=O)C(N)CC1=CC=C(O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)