accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

SOONOHO

CAS RN 40899-93-2

IUPAC Name: 1-(benzenesulfonyl)-1H-indole-2-carboxylic acid
Synonyms: 1-(benzenesulfonyl)indole-2-carboxylic acid
Molecular Weight (g/mol): 301.32
Molecular Formula: C15H11NO4S
InChi Key: QIWDUGKJAHJRAE-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC2=CC=CC=C2N1S(=O)(=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results